EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Occurrence of organophosphorus and carbamate pesticide residues in surface water samples from the Rangpur district of Bangladesh.
PMID 22526994
Abstract
We report the presence of organophosphorus and carbamate residues in 24 surface water samples and five ground water samples from Pirgacha Thana, Rangpur district, Bangladesh using high-performance liquid chromatography. A number of samples of surface water from paddy fields were found to contain chlorpyriphos, carbofuran and carbaryl at concentrations ranging from 0-1.189, 0-3.395 and 0-0.163 μg/L, respectively. Surface water from the lakes had chlorpyriphos, carbofuran and carbaryl at concentrations ranging from 0.544-0.895, 0.949-1.671 and 0-0.195 μg/L, respectively. This result indicates that the general public living in the area of Rangpur is at high risk of pesticide exposure from contaminated waters in the environment.