EMAIL THIS PAGE TO A FRIEND
AAPS PharmSciTech
Solvent removal and spore inactivation directly in dispensing vials with supercritical carbon dioxide and sterilant.
PMID 22528546
Abstract
A process is described using supercritical carbon dioxide to extract organic solvents from drug solutions contained in 30-mL serum vials. We report drying times of less than 1 h with quantitative recovery of sterile drug. A six-log reduction of three spore types used as biological indicators is achieved with direct addition of peracetic acid to a final concentration of approximately 5 mM (~0.04 %) to the drug solution in the vial. Analysis of two drugs, acetaminophen and paclitaxel, indicated no drug degradation as a result of the treatment. Furthermore, analysis of the processed drug substance showed that no residual peracetic acid could be detected in the final product. We have demonstrated an effective means to simultaneously dry and sterilize active pharmaceutical ingredients from organic solvents directly in a dispensing container.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
269336
Peracetic acid solution, 32 wt. % in dilute acetic acid
C2H4O3