EMAIL THIS PAGE TO A FRIEND
Journal of biochemistry
An anticancer agent, pyrvinium pamoate inhibits the NADH-fumarate reductase system--a unique mitochondrial energy metabolism in tumour microenvironments.
PMID 22528668
Abstract
Increased glycolysis is the principal explanation for how cancer cells generate energy in the absence of oxygen. However, in actual human tumour microenvironments, hypoxia is often associated with hypoglycemia because of the poor blood supply. Therefore, glycolysis cannot be the sole mechanism for the maintenance of the energy status in cancers. To understand energy metabolism in cancer cells under hypoxia-hypoglycemic conditions mimicking the tumour microenvironments, we examined the NADH-fumarate reductase (NADH-FR) system, which functions in parasites under hypoxic condition, as a candidate mechanism. In human cancer cells (DLD-1, Panc-1 and HepG2) cultured under hypoxic-hypoglycemic conditions, NADH-FR activity, which is composed of the activities of complex I (NADH-ubiquinone reductase) and the reverse reaction of complex II (quinol-FR), increased, whereas NADH-oxidase activity decreased. Pyrvinium pamoate (PP), which is an anthelmintic and has an anti-cancer effect within tumour-mimicking microenvironments, inhibited NADH-FR activities in both parasites and mammalian mitochondria. Moreover, PP increased the activity of complex II (succinate-ubiquinone reductase) in mitochondria from human cancer cells cultured under normoxia-normoglycemic conditions but not under hypoxia-hypoglycemic conditions. These results indicate that the NADH-FR system may be important for maintaining mitochondrial energy production in tumour microenvironments and suggest its potential use as a novel therapeutic target.