EMAIL THIS PAGE TO A FRIEND
Kardiologia polska
[Not so simple procedure in "simple" arrhythmias of ventricular].
PMID 22528708
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4670
Propafenone hydrochloride
C21H27NO3 · HCl