EMAIL THIS PAGE TO A FRIEND
Organic letters
Palladium-catalyzed asymmetric 6-endo cyclization of dienamides with substituent-driven activation.
PMID 22530559
Abstract
Chiral 2-piperidinone compounds with various C-6 substituents were successfully synthesized via a Pd-catalyzed asymmetric 6-endo cyclization of dienamides, which were evidently activated by both N-p-toluenesulfonyl and C-3 ester substituents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V209
δ-Valerolactam, 98%
C5H9NO