EMAIL THIS PAGE TO A FRIEND
Journal of travel medicine
Inoperable cerebral alveolar echinococcosis controlled with high dosages of albendazole adjusted with monitoring of blood levels.
PMID 22530831
Abstract
Cerebral alveolar echinococcosis (AE) is a rare and difficult-to-treat zoonosis caused by Echinococcus multilocularis. A 29-year-old immigrant from Siberia with a past history of hepatic AE, presented with acute onset of grand mal seizures, weakness of the left leg, and cephalgia. Magnetic resonance imaging of the brain revealed inoperable right-sided infiltrative lesions, suggesting cerebral AE. Despite anthelmintic treatment only slow improvement occurred.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35395
Albendazole sulfoxide, VETRANAL, analytical standard
C12H15N3O3S