EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
An efficient synthetic approach to 6,5'-(S)- and 6,5'-(R)-cyclouridine.
PMID 22531603
Abstract
Here we present new routes for the efficient syntheses of 6,5'-(S)- and 6,5'-(R)-cyclouridine. The syntheses utilize readily accessible uridine as a starting material. This route to the R diastereomer is significantly more efficient than previous synthetic efforts, allowing us to obtain large amounts of pure material for future biological testing.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
324442
O2,2′-Cyclouridine, 99%
C9H10N2O5