EMAIL THIS PAGE TO A FRIEND
Toxicology and applied pharmacology
Impact of environmental exposures on ovarian function and role of xenobiotic metabolism during ovotoxicity.
PMID 22531813
Abstract
The mammalian ovary is a heterogeneous organ and contains oocyte-containing follicles at varying stages of development. The most immature follicular stage, the primordial follicle, comprises the ovarian reserve and is a finite number, defined at the time of birth. Depletion of all follicles within the ovary leads to reproductive senescence, known as menopause. A number of chemical classes can destroy follicles, thus hastening entry into the menopausal state. The ovarian response to chemical exposure can determine the extent of ovotoxicity that occurs. Enzymes capable of bioactivating as well as detoxifying xenobiotics are expressed in the ovary and their impact on ovotoxicity has been partially characterized for trichloroethylene, 7,12-dimethylbenz[a]anthracene, and 4-vinylcyclohexene. This review will discuss those studies, as well as illustrate where knowledge gaps remain for chemicals that have also been established as ovotoxicants.