EMAIL THIS PAGE TO A FRIEND
Statistics in medicine
Handling the limit of detection by extrapolation.
PMID 22532151
Abstract
A general method of estimation with a variable observed subject to a limit of detection is introduced. It is based on extrapolation of the estimates obtained by increasing the limit of detection. Theoretical arguments support the method in some special cases, and it is explored by simulations. Several examples are presented and adaptations for other kinds of censoring explored.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O9389
Oltipraz, ≥98% (HPLC), powder
C8H6N2S3