EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Large increase of the lifetime of a charge-separated state in a molecular triad induced by hydrogen-bonding solvent.
PMID 22532315