EMAIL THIS PAGE TO A FRIEND
CNS neuroscience & therapeutics
Kallikrein gene transfer induces angiogenesis and further improves regional cerebral blood flow in the early period after cerebral ischemia/reperfusion in rats.
PMID 22533724
Abstract
The aims of this study were to find out whether kallikrein could induce angiogenesis and affect the cerebral blood flow (rCBF) in the early period after cerebral ischemia/reperfusion (CI/R). The adenovirus carried human tissue kallikrein (HTK) gene was administrated into the periinfarction region after CI/R. At 12, 24, and 72 h after treatments, neurological deficits were evaluated; expression of HTK and vascular endothelial growth factor (VEGF) were detected by immunohistochemistry staining; the infarction volume was measured; and rCBF was examined by( 14) C-iodoantipyrine microtracing technique. The expression of VEGF was enhanced significantly in pAdCMV-HTK group than controls over all time points (P < 0.05). Furthermore, the rCBF in pAdCMV-HTK group increased markedly than controls at 24 and 72 h after treatment (P < 0.05), and the improved neurological deficit was accompanied by reduced infarction volume in pAdCMV-HTK group 24 and 72 h posttreatment. In the early period after CI/R, kallikrein could induce the angiogenesis and improve rCBF in periinfarction region, and further reduce the infarction volume and improve the neurological deficits.