EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Preparation of SRN1-type coupling adducts from aliphatic gem-dinitro compounds in ionic liquids.
PMID 22534663
Abstract
S(RN)1-type coupling adducts are readily prepared by the reaction between a-sulfonylesters or a-cyanosulfones and gem-dinitro compounds in ionic liquids. The reactions progress smoothly and recovered ionic liquids can be used for several iterations, as long as they are washed with water to remove alkali metallic salts. The reaction rate is slower than the corresponding S(RN)1 reaction in DMSO, but no acceleration on irradiation or no inhibition in the presence of m-DNB are observed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
102369
1,4-Dinitrobenzene, 98%
C6H4N2O4
442250
1,4-Dinitrobenzene, analytical standard
C6H4N2O4