EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Diastereoselective synthesis of vicinal amino alcohols.
PMID 22535485
Abstract
The vicinal amino alcohol is a common motif in natural products and pharmaceuticals. Amino acids constitute a natural, inexpensive, and enantiopure choice of starting material for the synthesis of such functionalities. However, the matters concerning diastereoselectivity are not obvious. This Perspective takes a look in the field of diastereoselective synthesis of vicinal amino alcohols starting from amino acids using various methods.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
78260
Vicine, analytical standard
C10H16N4O7