Description
Molecular Formula
Add to Cart
D6140
Demeclocycline hydrochloride, powder or crystals
C21H21ClN2O8 · HCl
46161
Demeclocycline hydrochloride hydrate, VETRANAL, analytical standard
C21H21ClN2O8 · HCl · xH2O