EMAIL THIS PAGE TO A FRIEND
Anaerobe
Antimicrobial effect of linalool and α-terpineol against periodontopathic and cariogenic bacteria.
PMID 22537719
Abstract
Linalool and α-terpineol exhibited strong antimicrobial activity against periodontopathic and cariogenic bacteria. However, their concentration should be kept below 0.4 mg/ml if they are to be used as components of toothpaste or gargling solution. Moreover, other compounds with antimicrobial activity against periodontopathic and cariogenic bacteria should be used in combination.