EMAIL THIS PAGE TO A FRIEND
Electrophoresis
Analysis of alpha-1-acid glycoprotein isoforms using CE-LIF with fluorescent thiol derivatization.
PMID 22539314
Abstract
The analysis of glycoprotein isoforms is of high interest in the biomedical field and clinical chemistry. Many studies have demonstrated that some glycoprotein isoforms could serve as biomarkers for several major diseases, such as cancers and vascular diseases, among others. Capillary zone electrophoresis (CZE) is a well-established technique to separate glycoprotein isoforms, however, it suffers from limited sensitivity when UV-Vis detection is used. On the other hand, with laser-induced fluorescence (LIF) detection, derivatization reaction to render the proteins fluorescent can destroy the resolution of the isoforms. In this work, a derivatization procedure through the thiol groups of glycoproteins using either 5-(iodoacetamide) fluorescein (5-IAF) or BODIPY iodoacetamide is presented with the model protein of alpha-1-acid glycoprotein (AGP). The derivatization process presented enabled high-resolution analysis of AGP isoforms by CZE-LIF. The derivatization procedure was successfully applied to label AGP from samples of serum and secretome of artery tissue, enabling the separation of the AGP isoforms by CE-LIF in natural samples at different concentration levels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I9271
5-(Iodoacetamido)fluorescein, ≥90% (HPLC)
C22H14INO6