EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Immobilisation of quantum dots by bio-orthogonal PCR amplification and labelling for direct gene detection and quantitation.
PMID 22540122
Abstract
A sensitive and versatile detection scheme based on quantum dot immobilisation on a solid support through bio-orthogonal PCR amplification and labelling has been developed for detection and quantification of gene targets in complex DNA mixtures.