EMAIL THIS PAGE TO A FRIEND
Clinical nephrology
Maintenance therapy with single-daily, high-dose mizoribine after cyclophosphamide therapy for prepubertal boys with severe steroid-dependent nephrotic syndrome.
PMID 22541687
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M3047
Mizoribine, ≥98% (TLC)
C9H13N3O6