EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Efficient activation of thioglycosides with N-(p-methylphenylthio)-ε-caprolactam-TMSOTf.
PMID 22542575
Abstract
N-(p-Methylphenylthio)-ε-caprolactam (1) in combination with trimethylsilyl trifluoromethanesulfonate (TMSOTf) provides an efficient thiophilic promoter system, capable of activating different thioglycosides. Both 'armed' and 'disarmed' thioglycosyl donors were activated for glycosidic bond formation. Notably, this reagent combination works well in reactivity-based one-pot oligosaccharide assembly strategy.