EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Lysine-specific demethylase 1 (LSD1) and histone deacetylase 1 (HDAC1) synergistically repress proinflammatory cytokines and classical complement pathway components.
PMID 22542627