EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis of chalcone-amidobenzothiazole conjugates as antimitotic and apoptotic inducing agents.
PMID 22543234
Abstract
A series of chalcone-amidobenzothiazole conjugates (9a-k and 10a,b) have been synthesized and evaluated for their anticancer activity. All these compounds exhibited potent activity and the IC(50) of two potential compounds (9a and 9f) against different cancer cell lines are in the range of 0.85-3.3 μM. Flow cytometric analysis revealed that these compounds induced cell cycle arrest at G2/M phase in A549 cell line leading to caspase-3 dependent apoptotic cell death. The tubulin polymerization assay (IC(50) of 9a is 3.5 μM and 9f is 5.2 μM) and immuofluorescence analysis showed that these compounds effectively inhibit microtubule assembly at both molecular and cellular levels in A549 cells. Further, Annexin staining also suggested that these compounds induced cell death by apoptosis. Moreover, docking experiments have shown that they interact and bind efficiently with tubulin protein. Overall, the current study demonstrates that the synthesis of chalcone-amidobenzothiazole conjugates as promising anticancer agents with potent G2/M arrest and apoptotic-inducing activities via targeting tubulin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
101338
Benzothiazole, 96%
C7H5NS
W325600
Benzothiazole, ≥96%, FG
C7H5NS