EMAIL THIS PAGE TO A FRIEND
Journal of occupational and environmental medicine
Exposure estimates for workers in a facility expanding Libby vermiculite: updated values and comparison with original 1980 values.
PMID 22544162
Abstract
Low cumulative fiber exposure (CFE) has been associated with health effects in a cohort exposed to Libby vermiculite. This study refines the original 1980 exposure estimates and compares the CFE results. Cumulative fiber exposure estimates were developed using three times more industrial hygiene measurements and long-term workers' input. New adjustments included vermiculite ore source, seasonal overtime hours, time spent in various tasks, and recollection of historical dustiness. The overall mean (95% confidence interval) CFE (n = 513) in 1980 (0.80 [0.69 to 0.93]) was statistically similar to the overall mean (95% confidence interval) CFE in 2010 (0.74 [0.61 to 0.90]). The mean CFE in the lowest exposure category (<2 fiber-years/cm) decreased from 0.36 to 0.22 fiber-years/cm (P < 0.05). The 2010 CFE estimate extended the upper bound of the range of previous estimates from 28.10 to 106.31 fiber-years/cm. The range of CFE values was expanded. These estimates may impact the understanding of Libby vermiculite health outcomes.