EMAIL THIS PAGE TO A FRIEND
Environmental toxicology and chemistry
Aquatic toxicity tests with substances that are poorly soluble in water and consequences for environmental risk assessment.
PMID 22544669
Abstract
Aquatic toxicity tests with substances that are poorly soluble in water have been conducted using different methods, and estimates of toxicity have varied accordingly. The present study illustrates differences in toxicity values resulting from variation in test designs and solution preparation methods, and offers guidance on the best way to conduct these tests. Consequences for environmental risk assessment and classification are also discussed. The present study mainly considers active ingredients of plant protection products, but is also considered relevant to other chemicals. It is recommended that toxicity tests be conducted only up to the saturation limit, dispersants avoided, and solvents used only if necessary to support handling and speed of dissolution. Analytical measurements of exposure concentrations should reflect what organisms are exposed to. If acute toxicity testing at the saturation limit yields no adverse effects, further testing should not normally be required; the toxicity value of the endpoints should be considered as the saturation limit and adverse classification should not be required. Chronic testing, if required, should then be conducted at the practical saturation limit as this is the most realistic worst-case exposure scenario. If no adverse effects occur, the risk should be acceptable because higher aqueous exposure cannot occur. This could be substantiated by testing additional species. Assessment factors on no observed effect concentration (NOEC) values at the saturation limit require careful consideration in the risk assessment to avoid unnecessarily low regulatory acceptable concentrations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45751
Diflufenican, PESTANAL®, analytical standard
C19H11F5N2O2