EMAIL THIS PAGE TO A FRIEND
TheScientificWorldJournal
A facile synthesis of new 2-amino-4H-pyran-3-carbonitriles by a one-pot reaction of α, α'-bis(arylidene) cycloalkanones and malononitrile in the presence of K2CO3.
PMID 22545011
Abstract
A rapid and environmentally friendly method is developed for the synthesis of a series of new substituted 2-amino-4H-pyran-3-carbonitriles through a one-pot condensation of malononitrile and α, α'-bis(arylidene) cycloalkanones in ethanol by using K(2)CO(3) as a catalyst. Short experimental reaction times, excellent yields, no need to use cumbersome apparatus for purification of the products, and inexpensiveness and commercially availability of the catalyst are the advantages of this method.