EMAIL THIS PAGE TO A FRIEND
Nano letters
Hybrid multiferroic nanostructure with magnetic-dielectric coupling.
PMID 22545916
Abstract
The development of methods to economically synthesize single wire structured multiferroic systems with room temperature spin-charge coupling is expected to be important for building next-generation multifunctional devices with ultralow power consumption. We demonstrate the fabrication of a single nanowire multiferroic system, a new geometry, exhibiting room temperature magnetodielectric coupling. A coaxial nanotube/nanowire heterostructure of barium titanate (BaTiO(3), BTO) and cobalt (Co) has been synthesized using a template-assisted method. Room temperature ferromagnetism and ferroelectricity were exhibited by this coaxial system, indicating the coexistence of more than one ferroic interaction in this composite system.