EMAIL THIS PAGE TO A FRIEND
Asian Pacific journal of tropical medicine
Influence of Withania somnifera on obsessive compulsive disorder in mice.
PMID 22546655
Abstract
To study the influence of methanolic and aqueous extract of Withania somnifera (W. somnifera) root on the marble-burying behavior of mice a well-accepted model of obsessive compulsive behavior. Mice were divided in different groups (n = 6). Fluoxetine (5, 10, 15 mg/kg), (10, 25, 50, 100 mg/kg) and methanolic extract W. somnifera (MEWS) (10, 25, 50, 100 mg/kg) were administered i.p. 30 min. prior to the assessment of marble burying behavior and locomotor activity. The control group received vehicle of the extract. Administration of aqueous extracts W. somnifera (AEWS) and MEWS (50 mg/kg) successively decreased the marble burying behavior activity without affecting motor activity. This effect of AEWS and MEWS was comparable to standard fluoxetine, ritanserin and parachlorophenylalanine. W. somnifera extract is effective in treating obsessive compulsive disorder.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R103
Ritanserin, powder
C27H25F2N3OS