EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Wake promoting agents: search for next generation modafinil, lessons learned: part III.
PMID 22546675
Abstract
In searching for a next generation molecule to the novel wake promoting agent modafinil (compound 1), a series of fluorene-derived wakefulness enhancing agents were developed and evaluated in rat. Extensive pharmacokinetic studies of a potent member of the series (compound 15) revealed that the wake promotion activity of the analog was likely due to an active metabolite (compound 3).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H31204
9-Hydroxyfluorene, 96%
C13H10O