EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
In vitro chemoenzymatic and in vivo biocatalytic syntheses of new beauvericin analogues.
PMID 22547105
Abstract
New beauvericins have been synthesized using the nonribosomal peptide synthetase BbBEAS from the entomopathogenic fungus Beauveria bassiana. Chemical diversity was generated by in vitro chemoenzymatic and in vivo whole cell biocatalytic syntheses using either a B. bassiana mutant or an E. coli strain expressing the bbBeas gene.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B7510
Beauvericin, ≥97% (HPLC)
C45H57N3O9
37012
Beauvericin, VETRANAL, analytical standard
C45H57N3O9