EMAIL THIS PAGE TO A FRIEND
Hypertension (Dallas, Tex. : 1979)
Calcium antagonist added to angiotensin receptor blocker: a recipe for reducing blood pressure variability?: evidence from day-by-day home blood pressure monitoring.
PMID 22547446
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A7106
Azelnidipine, ≥98% (HPLC), powder
C33H34N4O6