EMAIL THIS PAGE TO A FRIEND
Endocrinology
A novel therapeutic approach to treating obesity through modulation of TGFβ signaling.
PMID 22549226
Abstract
Obesity results from disproportionately high energy intake relative to energy expenditure. Many therapeutic strategies have focused on the intake side of the equation, including pharmaceutical targeting of appetite and digestion. An alternative approach is to increase energy expenditure through physical activity or adaptive thermogenesis. A pharmacological way to increase muscle mass and hence exercise capacity is through inhibition of the activin receptor type IIB (ActRIIB). Muscle mass and strength is regulated, at least in part, by growth factors that signal via ActRIIB. Administration of a soluble ActRIIB protein comprised of a form of the extracellular domain of ActRIIB fused to a human Fc (ActRIIB-Fc) results in a substantial muscle mass increase in normal mice. However, ActRIIB is also present on and mediates the action of growth factors in adipose tissue, although the function of this system is poorly understood. In the current study, we report the effect of ActRIIB-Fc to suppress diet-induced obesity and linked metabolic dysfunctions in mice fed a high-fat diet. ActRIIB-Fc induced a brown fat-like thermogenic gene program in epididymal white fat, as shown by robustly increased expression of the thermogenic genes uncoupling protein 1 and peroxisomal proliferator-activated receptor-γ coactivator 1α. Finally, we identified multiple ligands capable of reducing thermogenesis that represent likely target ligands for the ActRIIB-Fc effects on the white fat depots. These data demonstrate that novel therapeutic ActRIIB-Fc improves obesity and obesity-linked metabolic disease by both increasing skeletal muscle mass and by inducing a gene program of thermogenesis in the white adipose tissues.