EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
An endoglycosidase with alternative glycan specificity allows broadened glycoprotein remodelling.
PMID 22551167
Abstract
Protein endoglycosidases are useful for biocatalytic alteration of glycans on protein surfaces, but the currently limited selectivity of endoglycosidases has prevented effective manipulation of certain N-linked glycans widely found in nature. Here we reveal that a bacterial endoglycosidase from Streptococcus pyogenes , EndoS, is complementary to other known endoglycosidases (EndoA, EndoH) used for current protein remodeling. It allows processing of complex-type N-linked glycans +/- core fucosylation but does not process oligomannose- or hybrid-type glycans. This biocatalytic activity now addresses previously refractory antibody glycoforms.