EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Determination of multiresidues in rapeseed, rapeseed oil, and rapeseed meal by acetonitrile extraction, low-temperature cleanup, and detection by liquid chromatography with tandem mass spectrometry.
PMID 22551191
Abstract
A multiresidue method for determining pesticides in rapeseed, rapeseed oil, and rapeseed meal by use of liquid chromatography-tandem mass spectrometry is developed. Samples were extracted with acetonitrile or acidified acetonitrile and cleaned up by a 12 h freezing step. The recovery data were obtained by spiking blank samples at three concentration levels. The recoveries of 27 selected pesticides in rapeseed, rapeseed oil, and rapeseed meal were in the range of 70-118%, at the concentration level of 10 μg kg(-1), with intraday and interday precisions of lower than 22 and 27%, respectively. Linearity was studied between 2 and 500 μg L(-1) with determination coefficients (R(2)) of higher than 0.98 for all compounds in the three matrices. The limits of quantitation (LOQs) of pesticides in rapeseed, rapeseed oil, and rapeseed meal ranged from 0.3 to 18 μg kg(-1). The n-octanol-water partition coefficient showed more influence than water solubility in extracting pesticides by acetonitrile from matrices of high fat content. This method was successfully applied for routine analysis in commercial products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
83450 Rapeseed oil from Brassica rapa, natural