EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
A Diels-Alder-based total synthesis of (-)-kainic acid.
PMID 22551444
Abstract
An efficient synthesis of (-)-kainic acid, through a high-pressure-promoted Diels-Alder cycloaddition of a vinylogous malonate derived from 4-hydroxyproline, is described. The bicyclic adduct could be converted into the natural product with complete stereocontrol.