EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Glycosylation of vanillin and 8-nordihydrocapsaicin by cultured Eucalyptus perriniana cells.
PMID 22552154
Abstract
Glycosylation of vanilloids such as vanillin and 8-nordihydrocapsaicin by cultured plant cells of Eucalyptus perriniana was studied. Vanillin was converted into vanillin 4-O-β-D-glucopyranoside, vanillyl alcohol, and 4-O-β-D-glucopyranosylvanillyl alcohol by E. perriniana cells. Incubation of cultured E. perriniana cells with 8-nordihydrocapsaicin gave 8-nordihydrocapsaicin 4-O-β-D-glucopyranoside and 8-nordihydrocapsaicin 4-O-β-D-gentiobioside.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
175536
4-Hydroxy-3-methoxybenzyl alcohol, 98%
C8H10O3
W373702
Vanillyl alcohol, ≥98%, FG
C8H10O3