EMAIL THIS PAGE TO A FRIEND
Magnetic resonance in chemistry : MRC
Slow relaxation of longitudinal multispin orders in weakly and strongly coupled two-spin systems.
PMID 22552892
Abstract
Longitudinal multispin order (LOMO) corresponds to a nonequilibrium population distribution in spin systems that exhibit scalar (J), dipolar, or quadrupolar coupling. We investigated the relaxation of longitudinal two-spin order (2-LOMO) in systems that had either weakly or strongly J-coupled spins. Our results indicated longer relaxation times for the 2-LOMO state compared with the corresponding longitudinal single-spin state (1-LOMO). Accessing nuclear spin states that have relaxation times longer than T(1), without the use of external contrast agents, is potentially useful for in vivo imaging and also for studying systems using dynamically hyperpolarized nuclear spins where longer life times are sought to increase the time available to study (bio)chemical events.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
01973
4-Aminobenzoic acid, analytical standard
C7H7NO2