EMAIL THIS PAGE TO A FRIEND
Biology of reproduction
Follicular life, death, and perpetuity.
PMID 22553219
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0300000
Testosterone propionate, European Pharmacopoeia (EP) Reference Standard
C22H32O3