EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Synthesis, spectral, thermal, X-ray single crystal of new RuCl₂(dppb)diamine complexes and their application in hydrogenation of Cinnamic aldehyde.
PMID 22554619
Abstract
The preparation of new three trans-[RuCl(2)(dppb)(N-N)] with mixed diamine (N-N) and 1,4-bis-(diphenylphosphino)butane (dppb) ligands, starting from RuCl(2)(PPh(3))(3) as precursor is presented. The complexes are characterized on the basis of elemental analysis, IR, (1)H, (13)C and (31)P{(1)H}NMR, FAB-MS, TG/DTA and single crystal X-ray diffraction studies. Complex (2L(1)) crystallizes in the monoclinic unit cells with the space group P2(1). The catalysts are evaluated for their Cinnamic aldehyde hydrogenation. The catalysts show excellent activity and selectivity for the unsaturated carbonyl compound under mild conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
208523
Ruthenium(III) chloride, Ru content 45-55%
Cl3Ru