EMAIL THIS PAGE TO A FRIEND
Archives of oral biology
Erosive-abrasive tissue loss in dentine under simulated bulimic conditions.
PMID 22554994
Abstract
The eroded organic dentine matrix is remarkably resistant to mechanical impacts. Additional brushing abrasion of eroded dentine has only limited influence on tissue loss. Digestive enzymes (e.g., pepsin, trypsin) that can reach the oral cavity during reflux or vomiting can partially degrade the matrix. This degradation may have an influence on both the stability of the matrix against mechanical forces and the susceptibility of eroded dentine to combined chemo-mechanical impacts. Both were investigated in the present study. Dentine samples of four groups were cyclically demineralised (6×2 min/day, 9 days) with an HCl-pepsin-solution (pH 1.6, 1.5mg/ml pepsin) and treated with a trypsin-solution (6×10 min/day, 2000 BAEE units/ml) directly afterwards. One group served as control; specimens of three groups were additionally brushed (2×15s/day) directly after the first and last trypsin treatment with forces of 200g, 300g, and 400g. Loss of demineralised and mineralised tissue was determined profilometrically. Additionally, an SEM analysis was performed. Loss of mineralised tissue (μm, mean±SD) was: 135.7±10.9 (control), 165.2±30.8 (200g), 168.0±16.3 (300g), and 174.9±17.1 (400g). Tissue loss was increased significantly (p≤0.001) by brushing independently of the force used (n.s. between brushed groups). SEM revealed that in all groups, the matrix was equally thinned through enzymatic activity, but it was still present as a continuous band. The results indicate that brushing of dentine after impact of acid and enzymes resulted in an increased tissue loss; however, the matrix persisted on the surface despite enzymatic treatment and brushing with forces of up to 400g.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
223484
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
CNa2O3
222321
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
CNa2O3
223530
Sodium carbonate, powder, ≥99.5%, ACS reagent
CNa2O3
S2127
Sodium carbonate, ReagentPlus®, ≥99.5%
CNa2O3
31432
Sodium carbonate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., anhydrous, ≥99.8% (calc. to the dried substance)
CNa2O3
13418
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
CNa2O3
900503
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
CNa2O3
900505
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
CNa2O3
900504
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
CNa2O3
71360
Sodium carbonate decahydrate, puriss. p.a., ≥99.0% (T)
CNa2O3 · 10H2O
13414
Sodium carbonate decahydrate, puriss., meets analytical specification of Ph. Eur., BP, FCC, E500, 99.5-100.5% (ex dried subst.)
CNa2O3 · 10H2O
577782
Sodium carbonate decahydrate, 99.999% trace metals basis
CNa2O3 · 10H2O
S4132
Sodium carbonate monohydrate, ReagentPlus®, ≥99.5%
CNa2O3 · H2O
13568
Sodium carbonate monohydrate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, 99.5-100.5% (ex dried subst.)
CNa2O3 · H2O
230952
Sodium carbonate monohydrate, ACS reagent, ≥99.5%
CNa2O3 · H2O