EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Synthesis of optically active dihydropyrans from asymmetric [4 + 2] cycloaddition of β,γ-unsaturated α-ketoesters with allenic esters.
PMID 22555871
Abstract
β-Isocupreidine (β-ICD) catalyzed asymmetric [4 + 2] cycloaddition of β,γ-unsaturated α-ketoesters with allenic esters afforded ester-substituted functionalized dihydropyran derivatives in high yields along with high enantioselectivities under mild conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
294985
Allene, ≥95%
C3H4