EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
The effects of notoginsenoside R₁ on the intestinal absorption of geniposide by the everted rat gut sac model.
PMID 22561891
Abstract
Geniposide is derived from Gardenia jasminoides Ellis (Rubiaceae). Its anti-inflammatory, antithrombotic effects as well as its preventive effect against ischemic stroke have been reported. Radix notoginseng (Chinese name tienchi or sanqi) is the dried root of Panax notoginseng (Burk.) F.H. Chen, an herb noted for its promotion of blood circulation, blood stasis removal and pain alleviation, and has been widely utilized for the prevention and treatment of microcirculatory disturbances in China and other Asian countries for many years. Notoginsenoside R₁ is an effective and structurally representative bioactive constituent of R. notoginseng. In our preliminary study, notoginsenoside R₁ was able significantly to improve the bioavailability of geniposide in beagle dogs, but the underlying mechanisms remain unknown. The present study aimed to investigate the intestinal kinetic absorptive characteristics of geniposide as well as the absorptive behavior influenced by the co-administration of notoginsenoside R₁ using an in vitro everted rat gut sac model. The results showed good linear correlation between the geniposide absorption in sac contents and the incubation time from 0 to 120 min. The concentration dependence showed a non-linear correlation between the geniposide absorption and the concentrations 0.356-1.424 mg/mL, the absorption was saturated about 1.424 mg/mL. Notoginsenoside R₁ at 0.1 and 0.2mg/mL concentrations was able significantly to enhance the absorption of geniposide (1.424 mg/mL) by 1.7- and 1.4-fold. Moreover, verapamil, a well-known P-glycoprotein inhibitor, was able significantly to elevate the absorption of geniposide 2.4-fold. Notoginsenoside R₁ influenced geniposide's absorption in a way similar to that of a P-glycoprotein inhibitor. In conclusion, notoginsenoside R₁ significantly enhances the intestinal absorption of geniposide. As for the mechanism underlying the improvement of geniposide's bioavailability, it is proposed that notoginsenoside R₁ was able to decrease the efflux transport of geniposide by P-glycoproteins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0153
Geniposide, ≥98% (HPLC)
C17H24O10
N3915
Notoginsenoside R1, ≥98% (HPLC)
C47H80O18