EMAIL THIS PAGE TO A FRIEND
Doklady. Biochemistry and biophysics
Dose dependences of lipid microviscosity of biological membranes induced by synthetic antioxidant potassium phenosan salt.
PMID 22562635