EMAIL THIS PAGE TO A FRIEND
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections
Accumulation of heat shock proteins and dehydrins in the needles of scotch pine at the early stage of the PS II photoinhibition during the autumn adaptation of plants to winter conditions.
PMID 22562683