EMAIL THIS PAGE TO A FRIEND
Pathology
Bilateral ovarian steroid cell tumours and massive macronodular adrenocortical disease in a patient with hereditary leiomyomatosis and renal cell cancer syndrome.
PMID 22565324