EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
miR-TRAP: a benchtop chemical biology strategy to identify microRNA targets.
PMID 22566243
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P8399
Psoralen, ≥99% (HPLC)
C11H6O3
73633
Psoralen, analytical standard
C11H6O3