EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Study on pharmacokinetics of danshensu sodium in danshen dripping solution in Beagles].
PMID 22568243
Abstract
To detect pharmacokinetics of Danshensu Sodium in Danshen dripping solution in Beagles. Danshen dripping solution was dripped intravenously into healthy Beagles at a dose of 10 mL x kg(-1). Their plasma samples were extracted with acetic ether, the blood concentrations were determined by HPLC method. Danshensu Sodium showed a good linear relationship within the range of 0.225-18.000 mg x L(-1), with the lowest detectable limit of 0.113 mg x L(-1). Its pharmacokinetic parameters were as follows: tmax was 30 min, Cmax was (9.5742 +/- 2.3715) mg x L(-1), t1/2 was (19.23 +/- 2.97) min, CL was (0.0127 +/- 0.0030) L x min(-1) x kg(-1), AUC(0-tn) was (474.954 +/- 95.483) mg x min x L(-1) and AUC(0-infinity) was (482.494 +/- 95.353) mg x min x L(-1). The accurate, stable and reliable blood concentration method shows a one-compartment mode of Danshensu Sodium in Beagles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0679
Danshensu sodium salt, ≥98% (HPLC)
C9H9O5 · Na