EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Parallel synthesis of 2-substituted 6-(5-oxo-1-phenylpyrrolidin-3-yl)pyrimidine-5-carboxamides.
PMID 22569416
Abstract
A library of 24 6-(5-oxo-1-phenylpyrrolidin-3-yl)pyrimidine-5-carboxamides 10{1,2; 1-12} was prepared by a parallel solution-phase approach. The synthesis comprises a five-step transformation of itaconic acid (11) into 1-methyl and 1-phenyl substituted 6-(5-oxo-1-phenylpyrrolidin-3-yl)pyrimidine-5-carboxylic acids 17{1,2} followed by parallel amidation of 17{1,2} with a series of 12 aliphatic amines 18{1-12} to afford the corresponding carboxamides 10 in good overall yields and in 80-100% purity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I29204
Itaconic acid, ≥99%
C5H6O4