EMAIL THIS PAGE TO A FRIEND
Journal of pediatric endocrinology & metabolism : JPEM
Intraoperative sonographic localization of a fractured Supprelin implant in a pediatric patient: a case report.
PMID 22570970
Abstract
The subdermal implant is an effective delivery system for the gonadotropin-releasing hormone analog histrelin in the treatment of central precocious puberty. In younger patients, implantation and replacement of the implant is performed under general anesthesia. This obviates the need for frequent intramuscular injections and has been shown to be preferable to patients and families. However, the implantation procedures have been complicated by fracture during surgical withdrawal. This paper discusses one such case and the intraoperative technique that led to effective withdrawal of the fractured implant.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L2761
[des-Gly10, D-His(Bzl)6]-LH-RH ethylamide, ≥97% (HPLC), powder
C66H86N18O12