EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Tetradentate Schiff base ligands and their complexes: synthesis, structural characterization, thermal, electrochemical and alkane oxidation.
PMID 22571942
Abstract
Three Schiff base ligands (H(2)L(1)-H(2)L(3)) with N(2)O(2) donor sites were synthesized by condensation of 1,5-diaminonapthalene with benzaldehyde derivatives. A series of Cu(II), Co(II), Ni(II), Mn(II) and Cr(III) complexes were prepared and characterized by spectroscopic and analytical methods. Thermal, electrochemical and alkane oxidation reactions of the ligands and their metal complexes were investigated. Extensive application of 1D ((1)H, (13)C NMR) and 2D (COSY, HETCOR, HMBC and TOSCY) NMR techniques were used to characterize the structures of the ligands and establish the (1)H and (13)C resonance assignments of the three ligands. Ligands H(2)L(1) and H(2)L(3) were obtained as single crystals from THF solution and characterized by X-ray diffraction. Both molecules are centrosymmetric and asymmetric unit contains one half of the molecule. Catalytic alkane oxidation reactions with the transition metal complexes investigated using cyclohexane and cyclooctane as substrates. The Cu(II) and Cr(III) complexes showed good catalytic activity in the oxidation of cyclohexane and cyclooctane to desired oxidized products. Electrochemical and thermal properties of the compounds were also investigated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
29598
Cyclooctane, analytical standard
C8H16
C109401
Cyclooctane, ≥99%
C8H16