EMAIL THIS PAGE TO A FRIEND
Cells Tissues Organs
Evidence of the TNF-? System in the Human Achilles Tendon: Expression of TNF-? and TNF Receptor at both Protein and mRNA Levels in the Tenocytes.
PMID 22572155