EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Sublethal effects of 2,2-dichloropropionic acid (dalapon) on Fossaria cubensis, intermediate host of the liver fluke, Fasciola hepatica.
PMID 2257331
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35562
Dalapon, PESTANAL®, analytical standard
C3H4Cl2O2